MicroPulser™ Electroporator, Bio-Rad
Supplier: Bio-Rad
1652100
100446-902EA 4900 USD
100446-902
MicroPulser™ Electroporator, Bio-Rad
Electroporation Systems Electroporators
The MicroPulser™ Electroporator provides safe and reproducible transformation of bacteria, yeast, and other microorganisms
  • One-button pulse delivery
  • Rapid charge time for fast sample handling
  • Arc quenching (ARQ) system significantly reduces arcing, protecting against sample loss
  • High-voltage 3,000 V capability improves efficiency in larger-volume cuvettes
  • Compact, space-saving design
  • Audible and visible pulse indicators
  • Time constant and actual volts displayed to monitor reproducibility
Manual programming allows voltage to be selected in a 200–3,000 V range, with 10 V precision, and allows pulse width to be selected in a 1.0–4.0 ms range with 0.1 ms precision.
DNA, RNA, siRNA, proteins, virions, nucleotides, carbohydrates, and dyes can be electroporated into cells. The electroporator has preset programs optimized for common bacteria and fungi, and the ability to manually program voltage and pulse width.
Certifications: Meets EN61000- 3-2 and EN61000-3-3 harmonic, flicker, and voltage fluctuation standards, FCC, Class A IEC 1010, C.