T100™ Thermal Cycler, Bio-Rad
Supplier: Bio-Rad
1861096
100446-370EA 9276.92 USD
100446-370
T100™ Thermal Cycler, Bio-Rad
Thermal Cyclers
The compact T100™ Thermal Cycler is a small footprint personal thermal cycler
  • Easy intuitive programming and use, with a color touch screen
  • Thermal gradient technology can optimize an assay in a single run
  • Store up to 500 protocols, and an unlimited number on USB drives
  • Built-in library of protocols including long PCR, fast PCR and RT-PCR
  • Small footprint
  • Fits in any laboratory
Thermal gradient technology permits quick reaction optimization in a single run by simultaneously testing eight different temperatures across a range of up to 25°C. The cycler has graphical programming and a library of standard PCR protocols. Easy protocol management with personalized folders or a USB flash drive, which can also be used for protocol transfer and backup.