ProSwift WCX-1S Protein Columns, Thermo Scientific
Supplier: Thermo Scientific
064295
10044-548EA 1957.3 USD
10044-548
ProSwift WCX-1S Protein Columns, Thermo Scientific
Chromatography Columns
Fast ion-exchange separation and protein purification is possible using the Thermo Scientific™ ProSwift ion exchange (IEX) columns.
  • Durable and robust, these columns exhibit stability
  • Reproducibility even after hundreds of runs
These monolith ion-exchange columns deliver the outstanding resolving power of nonporous media combined with the high capacity of conventional porous media. They offer excellent separations using fast gradients at low and high flow rates, which improve productivity.