ProSwift SAX-1S Protein Columns, Thermo Scientific
Supplier: Thermo Scientific
064293
10044-544EA 1957.86 USD
10044-544
ProSwift SAX-1S Protein Columns, Thermo Scientific
Chromatography Columns
Quickly purify proteins by ion-exchange using the quaternary amine stationary phase of ProSwift™ SAX-1S LC columns.
  • Durable and robust, these columns exhibit stability
  • Reproducibility even after hundreds of runs
These monolith ion-exchange columns deliver the outstanding resolving power of nonporous media combined with the high capacity of conventional porous media. They offer excellent separations using fast gradients at low and high flow rates, which improve productivity.