Niobium ICP Standard, Inorganic Ventures
Supplier: INORGANIC VENTURES MS
CGNB1-500ML
100360-624EA 146.97 USD
100360-624
Niobium ICP Standard, Inorganic Ventures
Standards Cationic Standards Niobium Standards
Certified reference material for Niobium, traceable to NIST.
One year expiration date (defined by supplier as the amount of time that it should remain in use after opening).