Thorium ICP Standard, Inorganic Ventures
Supplier: INORGANIC VENTURES MS
CGTH1-500ML
100360-316EA
120.62
USD
100360-316
Thorium ICP Standard, Inorganic Ventures
Standards
Cationic Standards
Thorium Standards
Certified reference material for Thorium, traceable to NIST.
One year expiration date (defined by supplier as the amount of time that it should remain in use after opening).
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)