Hybond™ Blotting Paper, Cytiva
Supplier: Cytiva
RPN6101M
10036-018EA 128.79 USD
10036-018
Hybond™ Blotting Paper, Cytiva
Blotting Paper
Research laboratory blotting paper for use in gel electrophoresis in all molecular biology and protein studies applications.
  • Recommended for use with all Hybond™ blotting membranes
  • High purity and uniformity ensures consistent and reproducible results
  • Pure Cellulose Grade, 0.39 mm thick paper with medium wet strength and smooth surface