Rubidium ICP Standard, Inorganic Ventures
Supplier: INORGANIC VENTURES MS
CGRB10-500ML
100359-782EA 326.31 USD
100359-782
Rubidium ICP Standard, Inorganic Ventures
Standards Cationic Standards Rubidium Standards
Certified reference material for Rubidium, traceable to NIST.
One year expiration date (defined by supplier as the amount of time that it should remain in use after opening).