BioFX® Alkaline Phosphatase/Horseradish Peroxidase Block, Surmodics
Supplier: Surmodics
APHP-0100-01
100359-236EA
95
USD
100359-236
BioFX® Alkaline Phosphatase/Horseradish Peroxidase Block, Surmodics
Assays
Immunological Assays
BioFX® Alkaline Phosphatase/Horseradish Peroxidase Block is supplied as a ready-to-use blocking reagent for immunohistochemistry samples. The solution blocks both alkaline phosphatase and peroxidase endogenous enzymatic activity.
- HRP and AP Blocker
- 3 year shelf life
- Ready to use formulation
Alkaline Phosphatase/Horseradish Peroxidase Block blocks the endogenous alkaline phosphatase and horseradish peroxidase present in many immunohistochemistry samples.