BioFX® BCIP/NBT Purple One Component AP Membrane Substrate, Surmodics IVD, Inc.
Supplier: Surmodics
BCID-1000-01 BCID-0100-01
100359-216EA 335 USD
100359-216 100359-214
BioFX® BCIP/NBT Purple One Component AP Membrane Substrate, Surmodics IVD, Inc.
Assays ELISAs
BCIP/NBT Purple One Component AP Membrane Substrate contains 5-bromo-4-chloro-3-indolylphosphate and nitroblue tetrazolium for use in immunblotting or other membrane applications.
  • Three year shelf life
  • Ready to use formulation
  • One-component
The BCIP/NBT substrate provides a dark purple precipitate upon reaction with alkaline phosphtase.
Certifications: ISO 13485:2016 and 9001:2015 Certified.