BioFX® Chemiluminescent Ultra Sensitive HRP Membrane Substrate, Surmodics IVD, Inc.
Supplier: Surmodics
CHMM-0060-2C
100359-204EA 265 USD
100359-204
BioFX® Chemiluminescent Ultra Sensitive HRP Membrane Substrate, Surmodics IVD, Inc.
Electrophoresis Reagents Western Blotting Reagents
BioFX® Chemiluminescent Ultra Sensitive HRP Membrane Substrate is a luminol based formulation for western blot applications.
  • Picogram to femtogram levels of sensitivity
  • High signal-to-noise ratios
Chemiluminescent substrates provide a higher degree of sensitivity than other substrates to enable lower limits of detection to be achieved in Western Blot assays. The increase in sensitivity requires more vigilance during the optimization process through the proper selection of the luminol substrate, antibody/antigen titers and blockers.
Surmodics provides multiple 2-component HRP chemiluminescent substrates with various intensities to allow the assay developer to choose the appropropriate substrate for the application.
Certifications: ISO 13485:2016 and 9001 certified.