BioFX® Chemiluminescent Ultra Sensitive HRP Microwell Substrate, Surmodics IVD, Inc.
Supplier: Surmodics
CHMI-0060-2C
100359-144EA 225 USD
100359-144
BioFX® Chemiluminescent Ultra Sensitive HRP Microwell Substrate, Surmodics IVD, Inc.
Electrophoresis Reagents Western Blotting Reagents
Chemiluminescent substrates provide a higher degree of sensitivity than other substrates to enable lower limits of detection to be achieved in Western Blot assays. The increase in sensitivity requires more vigilance during the optimization process through the proper selection of the luminol substrate, antibody/antigen titers and blockers.
  • Higher degree of sensitivity
  • Lower limits of detection
Surmodics provides multiple 2-component HRP chemiluminescent substrates with various intensities to allow the assay developer to choose the appropropriate substrate for the application.
Chemiluminescent substrates enables lower limits of detection to be achieved in ELISA assays. The increase in sensitivity requires more vigilance during the optimization process through the proper selection of the luminol substrate, antibody/antigen titers and blockers.
Certifications: ISO 13485:2016 and 9001:2015 certified.