Whatman™ Application-Specific Filter Papers, Grade 0048, Whatman products (Cytiva)
Supplier: Cytiva
10348903
10035-660PK 358.98 USD
10035-660
Whatman™ Application-Specific Filter Papers, Grade 0048, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
Filter mat made from a mixture of cellulose and polyester. This mat is used for optically testing baby food (artificial milk) for textile fibers.