Whatman™ Ashless Filter Aids, Whatman products (Cytiva)
Supplier: Cytiva
1703-050
10035-286PK 119.2 USD
10035-286
Whatman™ Ashless Filter Aids, Whatman products (Cytiva)
Filters Filter Paper Quantitative Filter Paper
Ashless filter aids enhance filtration speed by coagulating precipitates or suspensions to form a thick retentive 'prefilter' layer on top of normal filter paper.
  • Easily dispersible
  • Powder is of a purity similar to that of ashless quantitative papers
  • Maximum ash content is 0.015%
Ordering information: Supplied with two ended scoop for measuring 0.50 or 2.5 g quantities.