Whatman™ MV050 Series Vacuum Filtration Apparatus, Whatman products (Cytiva)
Supplier: Cytiva
10440020
10035-020PK 1580 USD
10035-020 10035-026
Whatman™ MV050 Series Vacuum Filtration Apparatus, Whatman products (Cytiva)
Flasks Filter Flasks
MV series vacuum filtration devices are made of stainless steel, which is especially suitable for microbiological applications.
The system can be used up to 200ºC, is autoclavable, and can be sterilized by dry heat up to 180ºC.