Whatman™ Grade GF 8 Glass Microfiber Filters with Binder, Whatman products (Cytiva)
Supplier: Cytiva
10370119 10370172 10370111
10034-834PK 131.14 USD
10034-834 10034-836 10034-838 76423-722
Whatman™ Grade GF 8 Glass Microfiber Filters with Binder, Whatman products (Cytiva)
Filters Glass Fiber Filters
These glass microfibre filters made from borosilicate glass are used for filtration of coarse particles. They offer faster filtration and a higher air flow rate compared to the similar grade GF 9 filters.
  • Inorganic binder for increased strength
  • Fast filtration speed
  • Can be used as a prefilter for dense samples
  • Function as air pollution filters for detecting furans, PCB, and other chemicals
With the added strength of an inorganic binder, grade GF 8 glass microfibre filters serve as air pollution filters by retaining a range of heavy particles found in vapor samples.