Whatman™ Application-Specific Filter Papers, Grade 3459, Whatman products (Cytiva)
Supplier: Cytiva
10316619
10034-742PK 312.24 USD
10034-742
Whatman™ Application-Specific Filter Papers, Grade 3459, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
Creped cellulose filter papers with good retentivity at a relatively high filtration speed. They are used for the clarifying filtration of dried beet pulp extracts or beet juice after the addition of lead acetate for subsequent polarimetric sugar determination. They are specifically designed for the Venema unit (lead acetate method).