Support Rings with Clamps, Chemglass
Supplier: Chemglass
CG-3057-10
10034-446EA 42.31 USD
10034-446
Support Rings with Clamps, Chemglass
Supports Flask Supports
Solid ring with built in clamp for attachment directly to support rod.
  • Nickel plated zinc alloy
  • Overall length: 180 mm