Thermometer Holder, Chemglass
Supplier: Chemglass
CG-3511-01
10034-322PK 34.45 USD
10034-322
Thermometer Holder, Chemglass
Thermometer Holders
The molded polypropylene thermometer holder clips onto beakers and other vessels and can hold a thermometer from OD 5 to 11 mm.