Glassware Cleaning Brushes, Foam Tips, Chemglass
Supplier: Chemglass
CG-1144-02
10034-254EA 21.62 USD
10034-254
Glassware Cleaning Brushes, Foam Tips, Chemglass
Brushes
Laboratory Cleaning Brushes feature polypropylene bristles with foam bristles at the tip.
  • Excellent for use with wide mouth bottles, graduated cylinders, and beakers
  • 2" brush tip width