3D Shakers/Rockers, Nutating Mixer, Chemglass Inc
Supplier: Chemglass
CLS-4029-100
10034-220EA 562.2 USD
10034-220
3D Shakers/Rockers, Nutating Mixer, Chemglass Inc
Shakers and Mixers Rotating/Rocking Shakers
This economical 3D shaker gently mixes liquid samples with a smooth motion that combines the actions of an orbital shaker and a rocker.
  • Small footprint
  • 3D motion provides thorough, gentle mixing
  • Dimpled mat holds tubes firmly in place