Cordless, Handheld UV Lamp, Chemglass
Supplier: Chemglass
CLS-1625-100 CLS-1625-102 CLS-1625-104
10034-170EA 514.97 USD
10034-170 10034-172 10034-174
Cordless, Handheld UV Lamp, Chemglass
Lights UV Lights, Lamps and Bulbs
UV lamps can be used for observing and imaging DNA and RNA gels
  • Reliable high intensity UV radiation
  • Portable, lightweight, and ergonomic
  • Single or dual wavelength models available
  • Charger and rechargeable batteries included
The ergonomic handle allows a safe grip, even with gloves. The safety switch prevents exposure of user to UV radiation by switching off the lamp if turned upside down.
They are essential for medical hygiene, bio-engineering, molecular biology, genetic studies and biological production, anti-counterfeit inspection, pharmaceutical production, criminal and forensic examination, and verification.