Creatinine Colorimetric Detection Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-907-030A
10033-564EA 299.75 USD
10033-564
Creatinine Colorimetric Detection Kit, Enzo Life Sciences
Colorimeters Comparator Kits
Low price, rapid creatinine detection kit for diabetes, cardiovascular and renal research.
For the quantitative determination of urinary creatinine from any species.
The Creatinine colorimetric detection kit utilizes a single-step liquid detection reagent that is safer and less time consuming than other assay methods. This kit is calibrated against the NIST standard and offers reproducible results with less than 6% inter- and intra-assay variation.
Ordering information: Includes Microtiter plate, Standard, Detection Reagent.