PROTEOSTAT® Protein Aggregation Standards, Enzo Life Sciences
Supplier: Enzo Life Sciences
51039-KP002
10033-526EA 315.01 USD
10033-526
PROTEOSTAT® Protein Aggregation Standards, Enzo Life Sciences
Assays Protein Assays
This kit is ideal for reliable and accurate quantification of protein aggregation in solution.
The ProteoStat® Protein Aggregation standards are ideal for reliable and accurate quantification of protein aggregation in solution. The standards are used to determine low levels of aggregated protein in a sample by comparing the assay response of a test sample to that of the standard curve; comprised of standards with known concentrations of aggregated IgG. Protein test samples and protein aggregation standards should be analyzed in the same manner, by mixing them with detection reagent and using a fluorescence microplate reader to measure the fluorescence intensity values. As with any protein assay, different protein aggregates will elicit slightly greater or lesser fluorescence intensity response based upon their inherent amino acid composition and sequence. It is recommended that a standard curve be prepared each time the ProteoStat® Protein Aggregation assay (Prod. No. ENZ-51023) is performed. The ProteoStat® Protein Aggregation standards contain stabilized, high-quality reference samples for generating an accurate standard curve. The standard curve is formulated from different amounts of aggregated IgG, combined with monomeric IgG. Once reconstituted, the samples contain 12.5%, 6.25%, 3.13%, 1.56%, 0.78%, 0.39%, 0.20% and 0% aggregated protein, while the total protein concentration of each standard is maintained at 1mg/ml. The total protein concentration of the IgG standard has been calibrated by direct comparison to an internal protein standard, in order to ensure lot-to-lot consistency and accuracy. Orthogonal characterization methods, including laser-based nanoparticle tracking and micro-flow imaging, have been employed to validate monomer integrity during manufacturing.
The standards are used to determine low levels of aggregated protein in a sample by comparing the assay response of a test sample to that of the standard curve; comprised of standards with known concentrations of aggregated IgG.
Ordering information: Includes A. ProteoStat® Control (0.0% aggregate), B. ProteoStat® Control (0.20% aggregate), C. ProteoStat® Control (0.39% aggregate), D. ProteoStat® Control (0.78% aggregate), E. ProteoStat® Control (1.56% aggregate), F. ProteoStat® Control (3.13% aggregate), G. ProteoStat® Control (6.25% aggregate), H. ProteoStat® Control (12.5% aggregate), Nuclease Free Water, 5 ml.