Lab Sinks, Self Rimming, One Bowl, V-1 Series, Eagle MHC
Supplier: Eagle Group
SR10A1V1 SR10B1V1 SR12A1V1 SR14A1V1 SRU14A1V1 SR14B1V1 SR14C1V1 SR14D1V1 SR16A1V1 SR16B1V1 SR16C1V1 SR18A1V1 SR19A1V1 SR19B1V1 SR22A1V1 SR24A1V1 SR24B1V1
10033-184EA 808.06 USD
10033-184 10033-186 10033-188 10033-190 10033-192 10033-194 10033-196 10033-198 10033-200 10033-202 10033-204 10033-206 10033-208 10033-210 10033-212 10033-214 10033-216
Lab Sinks, Self Rimming, One Bowl, V-1 Series, Eagle MHC
Furniture Casework
Heavy gauge type 304 series stainless steel coved bowls with large radius.
  • One compartment
  • Self rimming, deep drawn design
  • Features 1³/₈" hole for mixing faucet
1¹⁄₂" diameter basket drain in center of bowl. Sink can be mounted on top or bottom of countertop.
Ordering information: SRU sink (10033-192) features an upturn on sides and rear. Top-mount only.