VWR® 400DEL Digital Benchtop Homogenizer
Supplier: VWR International
10032-344
10032-344EA 13157.16 USD
10032-344
VWR® 400DEL Digital Benchtop Homogenizer
Homogenizers Benchtop Homogenizers
The VWR® 400DEL Digital Benchtop Homogenizer unit is the optimal balance of power and superior performance, combining a high-watt motor and advanced technology in digital speed controls to result in the most reliable and durable laboratory benchtop homogenizer unit.
  • Compact digital benchtop unit with 1305 high-watt motor
  • Motorized stand with extra-length stand height for large containers
  • Wide processing volume range, from 0.03 ml to 30L
  • Able to homogenize, emulsify, blend, and/or mix
  • VWR® Generator probe sold separately
With a sample volume range capability from micro-liters to large vessels, the unit demonstrates enormous processing versatility
This EL version provides Extra Length to the stand height for use with larger containers. Between its three Turn Speed Knob ability to adjust rpm in the hundreds (opposed to thousands), and a brighter LED display, to unit provides significantly more user-friendly control. Additional features include the Dual-Power Light Indicator for simpler error/problem identification, and the motorized UP/DOWN stand to provide effortless, accurate, and smooth motor positioning. Also included is a 25.4×22.8 cm (10×9") U-shaped base.
It is designed to homogenize, emulsify, blend, and/or mix organic and inorganic materials. Used in combination with VWR® generator probes ranging in diameter from 5 mm to 59 mm, it can process samples from 0.03 ml to 30L (higher sample volumes depend on vessel and longer length generator probes).
Certifications: CE marked.
Accessories information: Generator probes must be purchased with homogenizer units.
Ordering information: Supplied with a two-year manufacturer's warranty. VWR® Generator probe(s) are required for use with the VWR® 400DEL Homogenizer model and are sold separately (VWR® GenPack Adapter is required for VWR® GenPack Generator Probes).