VWR® 400D Digital Benchtop Homogenizer
Supplier: VWR International
VW-01-01400DS
10032-342EA 11577.43 USD
10032-342
VWR® 400D Digital Benchtop Homogenizer
Homogenizers Benchtop Homogenizers
The VWR® 400D Digital Benchtop Homogenizer unit is the optimal balance of power and superior performance, combining a high-watt motor and advanced technology in digital speed controls to result in the most reliable and durable laboratory benchtop homogenizer unit.
  • Compact digital benchtop unit with 1305 high-watt motor
  • Able to homogenize, emulsify, blend, and/or mix
  • Ideal for processing volumes from 0.03 mL to 30L
  • Motorized UP/DOWN stand
  • VWR® Generator probe sold separately
With a sample volume range capability from micro-liters to large vessels, the unit demonstrates enormous processing versatility
Between its three Turn Speed Knob ability to adjust rpm in the hundreds (opposed to thousands), and a brighter LED display, the unit provides significantly more user-friendly control. Additional features include a Dual-Power Light Indicator for simpler error/problem identification, and the motorized UP/DOWN stand to provide effortless, accurate, and smooth motor positioning. Also included is a 25.4x22.8 cm (10x9") U-shaped base.
It is designed to homogenize, emulsify, blend, and/or mix organic and inorganic materials. Used in combination with VWR® generator probes ranging in diameter from 5mm to 59mm, it can process samples from 0.03 mL to 30L (higher sample volumes depend on vessel and longer length generator probes).
Certifications: CE marked.
Accessories information: Generator probes must be purchased with homogenizer units.
Ordering information: Supplied with a two-year manufacturer's warranty. VWR® Generator probe(s) are required for use with the VWR® 400D Homogenizer model and are sold separately (VWR® GenPack Adapter is required for VWR® GenPack Generator Probes).