VWR® 200 Homogenizer
Supplier: VWR International
10032-336
10032-336EA 1262.33 USD
10032-336
VWR® 200 Homogenizer
Homogenizers Handheld Homogenizers
The VWR® 200 Homogenizer unit is able to homogenize samples with volumes as small as 0.03 mL, or as large as 1L (higher sample volumes dependent on viscosity).
  • Lightweight handheld homogenizer unit
  • For processing within various tube sizes to small beakers
  • High-torque motor with variable speed adjustment of 5000 to 33,000 rpm
  • Use handheld unit or post mount to a stand
  • VWR® Generator probe sold separately
The VWR® 200 Homogenizer unit has a 144 watt, high-torque motor with variable speed adjustment, allowing for homogenization in mere seconds and a maximum noise level of only 68dB
The unit is ideal for use with VWR® GenPack generator probes for low-cost, repeat homogenization of many samples without the risk of cross-contamination.
A seperate ON/OFF switch conveniently allows for the slide switch speed control to remain at an ideal homogenizing speed.
Certifications: CE marked.
Accessories information: Generator probes must be purchased with homogenizer units.
Ordering information: Supplied with a two-year manufacturer's warranty. VWR® Generator probe(s) are required for use with the VWR® 200 Homogenizer model and are sold separately (VWR® GenPack Adapter is required for VWR® GenPack Generator Probes). The VWR® 200 Homogenizer Stand Assembly is available as an optional accessory.