VWR® Premium GenPack Homogenizing Package
Supplier: VWR International
VWPK01200MPGXL
10032-330KT 3281.38 USD
10032-330
VWR® Premium GenPack Homogenizing Package
Homogenizers Handheld Homogenizers
The VWR® Premium GenPack Homogenizing Package is designed for homogenizing multiple samples quickly and easily.
  • Reusable 7mm diameter generator probe
  • Ideal for processing samples over a range of toughness
  • Stainless steel construction
The unit includes a 12-pack of reusable stainless steel probes, which offer significant advantages over plastic disposable generator probes/tips
This homogenizing package is very unique by its reusable 7mm diameter generator probe 12-pack manufactured from superior 316 stainless steel and PTFE, which allows for homogenizing of samples without cross-contamination and without the need to stop. Advantages over plastic disposable generator probes: can process more difficult samples including tough tissues, can be sterilized by any method, and it is more economical because of its ability to be reused an unlimited number of times.
Ideal for processing soft to the toughest of samples within tubes ranging from 1.5 to 15 mL, without risk of cross-contamination or the need for cleaning between samples.
Certifications: CE marked.
Ordering information: The VWR® Premium GenPack Homogenizing Package includes VWR® 200 Homogenizer unit, VWR® GenPack Adapter, and a 12-pack of Multi-Gen 7XL generator probes.