BTX Multi-Well Electroporation Plates, Harvard Apparatus
Supplier: Harvard Apparatus
450450-M
10031-730EA 498.66 USD
10031-730
BTX Multi-Well Electroporation Plates, Harvard Apparatus
Electroporation Plates
Run multiple samples in seconds for rapid optimization of electroporation variables and unsurpassed sample through-put.
  • Transfection of Mammalian Cells
  • Transformation of Bacteria and Yeast
  • siRNA and cDNA Library Screening
Complete user control over electroporation settings allows for a wide range of optimization flexibility
Each MOS plate consists of either 96 or 25 individual wells with integrated electrodes. The 96-well plate follows traditional 96-well format i.e., 12 columns of 8 wells each. The 25-well format has 5 columns of 5 wells each. Four different Multi-Well Plates are available: 96-well with 4 mm gap, 96-well with 2 mm gap, 25-well with 4 mm gap and 25-well with 2 mm gap.
Saves valuable lab time, moves experiments further faster, provides reliable results and requires no expensive reagents. Eliminates cross-contamination with patented integrated electrodes and plate sealer.