Anti-CD3 epsilon Armenian Hamster Monoclonal Antibody [clone: 145-2C11]

Supplier: Biolegend
100301 100302
100301-BLEA 30.33 USD
100301-BL 100302-BL
Anti-CD3 epsilon Armenian Hamster Monoclonal Antibody [clone: 145-2C11]
Antibodies
Purified anti-mouse CD3ε [145-2C11]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC, IHC-F, IF, IP, WB; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD3epsilon
Clonality: Monoclonal
Clone: 145-2C11
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR