SecureSeal™ Thermal Adhesive Sealing Film, Simport Scientific
Supplier: Simport Scientific
T329-6
10030-948PK 355.85 USD
10030-948
SecureSeal™ Thermal Adhesive Sealing Film, Simport Scientific
Seals Microplate Seals
SecureSeal™ Thermal Adhesive Sealing Film is designed for real time qPCR application and features a special pressure sensitive DMSO-resistant silicone adhesive embedded inside the film itself.
A polyester backing with end tabs enables easy positioning on the plate. This polyolefin film is also perfectly suited for use with raised rim plates, ensuring reliable sealing around each well. SecureSeal™ Thermal Adhesive Sealing Film offers the highest transparency, and has an operating temperature range of –40 to 120°C (–40 to 248°F). Certified RNase-, DNase-, and DNA-free.