Glycerol monostearate ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

31180150MG
100299-276EA 349.15 USD
100299-276
Glycerol monostearate ≥99%
Glycerol monostearate

Storage conditions: Store in a cool, dry place

Formula: C₂₁H₄₄O₅
MDL Number: MFCD00036186
CAS Number: 31566-31-1
UN: 12352100
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR