1-Monodecanoin ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 2,3-Dihydroxypropyl decanoate

31100150MG
100299-258EA 494.31 USD
100299-258
1-Monodecanoin ≥99%
1-Monodecanoin
Formula: C₁₃H₂₆O₄
MDL Number: MFCD00056656
CAS Number: 2277-23-8
UN: 12352100
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR