Methyl tricosanoate ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Tricosanoic acid methyl ester

202300100MG
100299-078EA 353.08 USD
100299-078
Methyl tricosanoate ≥99%
Methyl tricosanoate
Formula: C₂₄H₄₈O₂
MW: 368.64
MDL Number: MFCD00009349
CAS Number: 2433-97-8
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0529d8702aa02b7bed37aa023884cf48bd03eda&vid=c0529d8702aa02b7bed37aa023884cf48bd03eda&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR