Methyl laurate ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Dodecylic acid methyl ester, Methyl dodecanoate, Lauric acid methyl ester, Dodecanoic acid methyl ester

201201100MG
100299-070EA 325.62 USD
100299-070
Methyl laurate ≥99%
Methyl laurate

Store in a cool, dry place

Formula: C₁₃H₂₆O₂
MW: 214.35 g/mol
Boiling Pt: 266
Melting Pt: 4.5
MDL Number: MFCD00008966
CAS Number: 111-82-0
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40f51d2b73ae218be3ed5b8ab83a750d8efd1d21&vid=40f51d2b73ae218be3ed5b8ab83a750d8efd1d21&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR