Tricosanoic acid

Supplier: INDOFINE CHEMICAL COMPANY MS

102300100MG
100299-066EA 329.54 USD
100299-066
Tricosanoic acid
Tricosanoic acid

Store in a cool, dry place

Formula: C₂₃H₄₆O₂
MDL Number: MFCD00002808
CAS Number: 2433-96-7
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d23e728210af260c414c20985019d56babeac19&vid=4d23e728210af260c414c20985019d56babeac19&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR