3-Fluoro-4-methyl-DL-phenylglycine

Supplier: INDOFINE CHEMICAL COMPANY MS

2637611GM
100298-554EA 319.08 USD
100298-554
3-Fluoro-4-methyl-DL-phenylglycine
3-Fluoro-4-methyl-DL-phenylglycine
Formula: C₉H₁₀FNO₂
MW: 183.18 g/mol
MDL Number: MFCD06247587
CAS Number: 261951-76-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bac256946876b6f6ccd083b17672a2a3b93be175&vid=bac256946876b6f6ccd083b17672a2a3b93be175&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR