2,3,4-Trifluoroaniline ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

261845100GM
100298-372EA 119 USD
100298-372
2,3,4-Trifluoroaniline ≥98%
2,3,4-Trifluoroaniline
Formula: C₆H₄F₃N
MDL Number: MFCD00011737
CAS Number: 3862-73-5
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58d29664e52d54f3ce4756aa20d819d4eba6bfcf&vid=58d29664e52d54f3ce4756aa20d819d4eba6bfcf&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR