Boc-D-Asn(Trt)-OH ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

049001GM
100297-928EA 228.85 USD
100297-928
Boc-D-Asn(Trt)-OH ≥98%
Boc-D-Asn(Trt)-OH
Formula: C₂₈H₃₀N₂O₅
MW: 474.6 g/mol
MDL Number: MFCD00153300
CAS Number: 210529-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR