D-(+)-Cellobiose ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: β-D-Glc-(1→4)-D-Glc, 4-β-D-glucopyranosyl-D-glueopyranose, Cellose, β-Cellobiose, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, 4-O-β-D-Glucopyranosyl-D-glucopyranose

04145925GM
100297-578EA 232.77 USD
100297-578
D-(+)-Cellobiose ≥98%, powder
D(+)-Cellobiose

  • White powder
  • Store in a cool, dry place

Formula: C₁₂H₂₂O₁₁
MW: 342.30 g/mol
Melting Pt: 235…245 °C
MDL Number: MFCD00136034
CAS Number: 528-50-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74d664aadf26994d8de796202822d152c3acf810&vid=74d664aadf26994d8de796202822d152c3acf810&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR