Fmoc-D-Arg(Mtr)-OH ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Nα-Fmoc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulfonyl)-D-arginine, Nα-Fmoc-Nomega-(4-methoxy-2,3,6-trimethylphenylsulphonyl)-D-arginine

0411951GM
100297-528EA 154.31 USD
100297-528
Fmoc-D-Arg(Mtr)-OH ≥99%
Fmoc-D-Arg(Mtr)-OH
Formula: C₃₁H₃₆N₄O₇S
MW: 608.71 g/mol
Melting Pt: 123…130 °C
MDL Number: MFCD00065616
CAS Number: 120075-24-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR