2,5-Dibromo-3,4,6-trifluoroaniline ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

270971GM
100297-460EA 281.15 USD
100297-460
2,5-Dibromo-3,4,6-trifluoroaniline ≥97%
2,5-Dibromo-3,4,6-trifluoroaniline
Formula: C₆H₂Br₂F₃N
MDL Number: MFCD01862003
CAS Number: 232267-32-2
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51d66c68f7fcd24f514a1858aa091a3e88333cb9&vid=51d66c68f7fcd24f514a1858aa091a3e88333cb9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR