4-Chloro-5,6-dimethyl-2-(trifluoromethyl)pyrimidine ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

263791GM
100297-330EA 432.85 USD
100297-330
4-Chloro-5,6-dimethyl-2-(trifluoromethyl)pyrimidine ≥97%
4-Chloro-5,6-dimethyl-2-(trifluoromethyl)pyrimidine
Formula: C₇H₆ClF₃N₂
MDL Number: MFCD00126659
CAS Number: 175277-32-4
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=588d243a6f9c5cd2332ad4ef4c35324964f3806e&vid=588d243a6f9c5cd2332ad4ef4c35324964f3806e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR