2',4',5'-Trifluoroacetophenone ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 1-(2,4,5-Trifluorophenyl)ethan-1-one, 2,4,5-Trifluoroacetophenone

0938195GM
100297-030EA 109.85 USD
100297-030
2',4',5'-Trifluoroacetophenone ≥99%
2',4',5'-Trifluoroacetophenone
Formula: C₈H₅F₃O
MW: 174.12
Boiling Pt: 76
Density: 1.38
MDL Number: MFCD00061193
CAS Number: 129322-83-4
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5a089eabc2ae8c690efa03342a0f80390213be7&vid=e5a089eabc2ae8c690efa03342a0f80390213be7&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29d216579dc24d5dc31fa83ce10e87daff246d08&vid=29d216579dc24d5dc31fa83ce10e87daff246d08&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR