4'-Cyano-2'-(trifluoromethoxy)acetanilide ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

093211GM
100296-972EA 143.85 USD
100296-972
4'-Cyano-2'-(trifluoromethoxy)acetanilide ≥97%
4'-Cyano-2'-(trifluoromethoxy)acetanilide
Formula: C₁₀H₇F₃N₂O₂
Melting Pt: 118…120 °C
MDL Number: MFCD00204174
CAS Number: 175278-19-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d2823138e1d84388f67fe86fcdbbd558d9f140e&vid=4d2823138e1d84388f67fe86fcdbbd558d9f140e&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50dfe7ee03a643bc65a3f5b4967a656f0396b7d5&vid=50dfe7ee03a643bc65a3f5b4967a656f0396b7d5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR