1-(4-(Trifluoromethyl)phenyl)imidazole ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

0921601GM
100296-932EA 176.54 USD
100296-932
1-(4-(Trifluoromethyl)phenyl)imidazole ≥98%
1-(4-(Trifluoromethyl)phenyl)imidazole
Formula: C₁₀H₇F₃N₂
Melting Pt: 71…73 °C
MDL Number: MFCD00060493
CAS Number: 25371-98-6
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae8f49af143314eab030c082cf22d0e453fcc422&vid=ae8f49af143314eab030c082cf22d0e453fcc422&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7294e627ac0adcdb29b6daa576b0d85d02182c4a&vid=7294e627ac0adcdb29b6daa576b0d85d02182c4a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR