1H,1H-Pentafluoropropyl methacrylate ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 1H,1H-Pentafluoropropyl methacrylate, 2,2,3,3,3-Pentafluoropropyl Methacrylate, Methacrylic acid 2,2,2,3,3-pentafluoropropyl ester, Methacrylic acid 2,2,3,3,3-Pentafluoropropyl ester, 1H,1H-Perfluoropropyl methacrylate

0918435GM
100296-912EA 146.46 USD
100296-912
1H,1H-Pentafluoropropyl methacrylate ≥97%
1H,1H-Pentafluoropropyl methacrylate
Formula: C₇H₇F₅O₂
MW: 218.12
MDL Number: MFCD00039256
CAS Number: 45115-53-5
UN: 3272
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d80430a3be3f00dccf00f22c636f34af19c510c&vid=7d80430a3be3f00dccf00f22c636f34af19c510c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR