Methyl-2-fluorobenzoate ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

09164525GM
100296-904EA 104.62 USD
100296-904
Methyl-2-fluorobenzoate ≥98%
Methyl 2-fluorobenzoate
Formula: C₈H₇FO₂
MDL Number: MFCD00017913
CAS Number: 394-35-4
UN: 12352100
ADR: ,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8fd380b96e6f637b3d3d4768321cb73195cf889e&vid=8fd380b96e6f637b3d3d4768321cb73195cf889e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR