4-Iodo-D-phenylalanine ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: H-D-Phe(4-I)-OH

0418381GM
100296-588EA 198.77 USD
100296-588
4-Iodo-D-phenylalanine ≥99%
4-Iodo-D-phenylalanine
Formula: C₉H₁₀INO₂
MW: 291.1 g/mol
MDL Number: MFCD00063067
CAS Number: 62561-75-5
UN: 12352100
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=019e24f94ff04bd439d133e56a6c06c9a97b093e&vid=019e24f94ff04bd439d133e56a6c06c9a97b093e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR